Would you like to log in again?
BRAND® Filtered Pipette Tips, 1250 µl
Supplier: BrandTech
BRAND®
732736 732738
77322-348PK 256.5 USD InStock
77322-348 77322-350
BRAND® Filtered Pipette Tips, 1250 µl
Pipette Tips
Produced and processed under cleanroom conditions to ensure contamination-free analysis.
  • Perfect fit for Transferpette® micropipettes and suitable for other popular pipette brands
  • Made from high-purity, additive-free PP
  • Manufactured without lubricants and cadmium-free pigments; no parting agents or demolding aides are used
  • PE filters are free from chemical additives
BRAND® pipette tips offer proven functionality for reliable and precise results. Tips are colorless and thin-walled, with visible graduations. Tips and TipBoxes are autoclavable at 121 °C.
Tips are neatly packaged in a sturdy box with a hinged lid for added convenience. Tips come in a distinctive purple-colored tip tray for easy recognition.
All tips are manufactured under control room conditions with intensive in-process controls, including batch management and final product inspection.
BIO-CERT® LIQUID HANDLING QUALITY tips are DNA-, RNase-, DNase-, and pyrogen-free. They are also free of PCR-inhibitors and ATP. Sterile tips are sterilized with beta radiation and are supplied with a Certificate of Analysis.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty