Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Terpinolene (p-mentha-1,4(8)-diene), natural isolate

Supplier: Ricca Chemical
Danger
Synonyms: Terpinolene

RCCT9002-20C6 RCCT9002-750C6 RCCT9002-100C6
RCCT9002-100C6EA 58.27 USD InStock
RCCT9002-100C6 RCCT9002-750C6 RCCT9002-20C6
Terpinolene (p-mentha-1,4(8)-diene), natural isolate
Terpinolene (p-Mentha-1,4(8)-diene)


Formula: C₁₀H₁₆
MDL Number: MFCD00049191
CAS Number: 586-62-9
UN: 2541
ADR: 3,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty