Would you like to log in again?
Uvex Avatar™ Blue Blocker Safety Eyewear
Supplier: Honeywell Safety
Uvex Avatar™
S2855HS
76637-526BX 345.44 USD InStock
76637-526
Uvex Avatar™ Blue Blocker Safety Eyewear
Safety Glasses
Stylish and protective Uvex Avatar™ eyewear has premium anti-fog coating and anti-scratch hard coating, and features technology allowing customizability. The eyewear also features nine base wraparound, indirect venting, wire-core temples, and MMT-MULTI MATERIAL TECHNOLOGY® at every touchpoint for maximum all-day comfort and a customizable fit for every facial profile.
  • PC lens: HydroShield™ anti-fog and hardcoat
  • Ratchet: Wire-core, adjustable temples, flexible nose, MMT
  • Indirect venting system: soft brow frame with MMT
  • Available in four modern frame colors and five lens tints
  • ³/₄ frame: Contemporary stylish fit for high performance
Extended exposure to high energy, visible blue-violet light (380 to 450 nm) can cause eye fatigue, headaches, mood changes, and sleep rhythm disturbances such as insomnia. The sources of this exposure are the sun, computers, handheld devices, and LED lighting.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty