Would you like to log in again?
Captia™ Measles IgM Kit
Supplier: Trinity Biotech
2326060
77441-874KT 2805.88 USD InStock
77441-874
Captia™ Measles IgM Kit
Assays ELISAs
Captia Measles IgM Assay (ELISA) is intended for the in vitro determination of Measles IgM antibodies in human serum.
  • RTU Reagents
  • Color Coded Reagents & Caps
  • Comprehensive Product insert for Ease of Use
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty