Would you like to log in again?
Recombinant Coronavirus (COVID-19 NP) Antigen (aa 2-419) (from E. coli)
Supplier: Enzo Life Sciences
PRT282-0100
76418-066EA 779.88 USD InStock
76418-066
Recombinant Coronavirus (COVID-19 NP) Antigen (aa 2-419) (from E. coli)
Proteins and Peptides
Produced in E. coli. Coronavirus (COVID-19) nucleoprotein (aa 2-419) is fused to a His-tag.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty