Would you like to log in again?
Stilla® naica® IQ/OQ Kit
Supplier: Stilla Technologies
Stilla®
R30001
77392-294EA 480 USD InStock
77392-294
Stilla® naica® IQ/OQ Kit
Nucleic Acid Reagents End-point PCR Enzymes and Kits
Routine Operational Qualification (OQ) procedures of the naica® system and Nio™+ can be performed using the naica® IQ/OQ Kit.
  • Ready-to-use OQ solution
  • 24 reactions on the sapphire chip (Two full OQ of the naica® system)
  • 96 ruby chip reactions (1 full OQ of the Nio™+)
The naica® IQ/OQ Kit is a ready-to-use OQ solution to closely monitor equipment compliance to operating specifications. Use the naica® IQ/OQ Kit to qualify the naica® system and Nio™+ on a periodic basis in accordance with established operating procedures (SOPs). Experience the streamlined routine OQ solution with the naica® IQ/OQ Kit for a wide range of key nucleic detection and quantification applications, validated for use on all Stilla Technologies dPCR systems.
Delivery information: Dry-ice shipment.
Caution: Store at −20 ±5 °C. For research use only. Not for use in diagnostic procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty