Would you like to log in again?
Dengue Virus Reporter Virus Particle (RVP) with Renilla Luciferase Reporter (Type 1)
Supplier: ReVacc Scientific
N91Y8-1ML N91Y8-200UL
77394-700EA 615 USD InStock
77394-700 77394-698
Dengue Virus Reporter Virus Particle (RVP) with Renilla Luciferase Reporter (Type 1)
Vectors, Plasmids and Libraries
Reporter virus particles (RVPs) package the capsid-preMembrane-envelope (C-prM-E) gene construct of Dengue virus (DENV) (GenBank: AAB70694.1, SEROTYPE 1 strain) and a Renilla luciferase reporter. As the wild typed Dengue virus is handled under BSL-2 precautions, this Dengue virus RVP can be handled using BSL-2 containment practices.
  • Applications:
  • Anti-Dengue virus neutralizing antibody screening at high throughput
  • Anti-Dengue virus drug screening at high throughput
  • Dengue virus vaccine efficacy evaluation at high throughput
  • Dengue virus pseudovirus transduction of target cells for viral entry, receptor recognition, cellular tropism and functional studies, such as ADCC or ADE analysis
Description: Infection of cells with this RVP carrying Renilla luciferase reporter results in high level Renilla luciferase activity. See our titration result showing that the starting 2-fold diluted RVP-infected target cells generates signal ~ 600-fold higher than uninfected control (cell alone as background in blue). We also evaluated the neutralizing activity of monoclonal antibody (mAb)-domain Ⅲ specific RV25.
Biosafety level: Handle it in biosafety cabinet in BSL-2. Contacted tips and tubes should be decontaminated by 10% disinfecting bleach.
Use protocol: A complete protocol is supplied with the shipping of product. Briefly, seed target cells (Vero or others expressing receptor). Next day, incubate 50 μl diluted virus with 50 μl your sample in each well of 96-well plate with at least one duplicate for 30 to 60 min at 37 °C. Then transfer the mixture (100 μl) to plate with cells pre-seeded. Read Renilla luciferase signal next day.
Accessories information: See suggested protocol to use this product.
Packaging: Shipping with dry ice. Require −80 °C storage. Multiple freeze/Thaw cycles will reduce its sensitivity. Recommend only 1 cycle. Aliquot after the first thaw.
Caution: Pseudoviruses are intended for research use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty