Would you like to log in again?
EZ-Accu Shot™ Burkholderia cenocepacia derived from ATCC® BAA-245™, Microbiologics
Supplier: MICROBIOLOGICS, INC.
EZ-Accu Shot™
01269A
76407-430EA 416.18 USD InStock
76407-430
EZ-Accu Shot™ Burkholderia cenocepacia derived from ATCC® BAA-245™, Microbiologics
Control reagents
Quantitative microorganism preparation that delivers less than 100 CFU per 0.1 ml of hydrated suspension.
  • No dilutions required
Kit contains: 5 pellets of a single enumerated strain and 5 vials of hydrating fluid (1.2 ml per vial).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty