Would you like to log in again?
miniPCR® blueGel™ Electrophoresis Classroom Startup Pack
Supplier: AMPLYUS LLC
miniPCR® blueGel™
QP-1500-44
470355-090EA 2005.99 USD InStock
470355-090
miniPCR® blueGel™ Electrophoresis Classroom Startup Pack
Educational Classroom Kits and Activities
This startup pack provides electrophoresis equipment for up to eight laboratory groups.
  • One-button operation
  • Observe band separation within 5 minutes
  • Real-time band visualization while running your gel
  • Safe nucleic acid staining with GelGreen® or SYBR-family dyes
  • Safe blue light illumination
  • Low voltage and automatic current shut-off when cover is removed
  • Direct photo documentation with included Fold-a-View™ imaging hood
Equip your laboratory or classroom to perform gel electrophoresis labs! The blueGel™ electrophoresis startup pack provides four blueGel™ electrophoresis systems and eight 20 L micropipettes - enough equipment for up to eight lab groups. The blueGel™ system is safe, fast, compact, durable, and easy to use. By integrating gel electrophoresis and blue light transillumination into an all-in-one device, you can see results instantly while still running your gel. There is no need for a separate imaging system, time-consuming staining and destaining, or harmful ethidium bromide and UV light. And with eight micropipettes, students will have all the equipment they need to load and run gels.
Certifications: UL and CE marked.
Ordering information: Backed by a 2-year warranty.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty