Would you like to log in again?
Filter Forceps, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
51147 51147 4690 4690
30033-042EA 51.22 USD InStock
30033-042 CA30033-042 34181-102 CA34181-102
Filter Forceps, Cytiva (Formerly Pall Lab)
Forceps
For easy placement of membrane filters into filter funnels or petri dishes.
  • Flat surface
  • Avoid membrane damage
  • Stainless steel
Tips have a flat, smooth surface to avoid membrane filter damage. Stainless steel construction with polypropylene finger grips for a comfortable and secure hold. Autoclavable.
Order Now
ORDER
COMPLEMENTARY PRODUCTS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty