Would you like to log in again?
Eight-Well PCR Tube Strip Adapters
Supplier: Azenta Life Sciences
4TI-0370 4TI-0371 4TI-0292
76638-134CS 187.18 USD InStock
76638-134 76637-962 76638-108
Eight-Well PCR Tube Strip Adapters
Racks Tube Racks
A 96-format frame that fits 8-well PCR tube strips with PC frame perfectly to allow for easy handling and use with automation.
  • Locator pins on the deck ensure the 8-well PCR tube strips with PC frame are always loaded in the correct orientation
  • A dedicated lid with interference fit provides protection for the strips during handling and shipping
  • Unlike heavy and solid adapters on the market, the 8-well PCR tube strip adapter can be affordably included in kits that are assembled and shipped to customer
  • Slightly taller than standard profile PCR plates, fitting perfectly standard profile PCR tubes without taking up excessive space
  • Dedicate lid: Certified free of DNase, RNase, human DNA, bacterial and eukaryotic cells, dust and endotoxins/pyrogens
  • Low profile, no cut corner, no condensation rings, clear polystyrene
Standard profile 8-well PCR tube strips with PC frame have an interference fit with the adapter, ensuring they are held within the plate and do not move around/fall out during handling.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty