Would you like to log in again?
Kleenex® Reveal™ Ultra Moisturizing Foam Hand Sanitizer, Kimberly-Clark
Supplier: Kimberly-Clark
Kleenex® Reveal™
45826
76333-052CS 150.74 USD InStock
76333-052
Kleenex® Reveal™ Ultra Moisturizing Foam Hand Sanitizer, Kimberly-Clark
Skincare Skin Disinfectants Liquid and Foam Skin Disinfectants
This solution was formulated to meet the CDC guidelines for alcohol-based sanitizers and helps stop the spread of germs by effectively killing up to 99.9% of bacteria, all without the use of water.
  • Luxurious formulation enriched with cucumber and aloe vera
  • Gentle for frequent use
  • Clear, Unscented
  • Perfect for maintaining hand hygiene while on the go
Kleenex® hand sanitizer is a popular choice for hotels, spas, and more. Because the sanitizer is NSF E-3 rated, it is also suitable for use in food processing and food service settings. Kleenex® Ultra Moisturizing Foam Hand Sanitizer adds a touch of luxury to hand hygiene and facility maintenance, making it easy for staff and guests to stay healthy.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty