Would you like to log in again?
Multi-MicroPlate Genie® Mixer Shaker, 230 V
Supplier: Scientific Industries
SI-4001A
89202-336EA 1564.93 USD InStock
89202-336
Multi-MicroPlate Genie® Mixer Shaker, 230 V
Shakers and Mixers Microplate Shakers
This mixer available in analog or digital models holds four microplates, and has an aggressive speed for increased mixing efficiency in every well
  • 230 V model
  • Analog or digital
  • Aggressive speed
  • Small orbit
  • True vortexing action
The Multi-MicroPlate Genie® accepts most SBS microplate types, including 384-well plates. The mixer's high speed and small orbit combine to offer true vortexing action in each well of the microplate for complete and uniform mixing. A built-in timer operates continuously or from 1 to 60 minutes for analog model, and to 99 minutes for digital model. Digital model incorporates pulsing feature.
Certifications: CE marked.
Accessories information: An accessory tray to increase capacity up to 8 microplates is available.
Ordering information: The 230 V model is supplied without a plug.
Caution: 230 V model. Corresponding model with voltage suitable for use in US (89202-334) is also available, please see separate product page.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty