Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Balance Blocker™ Tris

Supplier: Arlington Scientific
Balance Blocker™
PP91T-500
77395-202EA 110 USD InStock
77395-202
Balance Blocker™ Tris
Buffers Electrophoresis Buffers Blocking Buffers
A universal blocking reagent supplied in tris for diagnostic and research systems.

  • Contains a unique blend of non-mammalian proteins
  • Provides superior blocking in super sensitive assay systems
  • Supplied in HDPE plastic

Preservative - 0.09% sodium azide.

Applications: ELISA/WB/Lateral flow.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty