Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Aquatic Block™ PBS

Supplier: Arlington Scientific
Aquatic Block™
PP81P-500 PP81P-125 PP81P-1L
77395-174EA 110 USD InStock
77395-174 77395-172 77395-170
Aquatic Block™ PBS
Buffers Electrophoresis Buffers Blocking Buffers
A non-mammalian blocking reagent supplied in PBS for diagnostic systems involving membrane or other solid phase material.

  • Optimized for superior performance in super sensitive assay systems
  • Utilizes proteins that do not interact with mammalian antibodies
  • Supplied in HDPE plastic

Preservative - 0.09% sodium azide.

Applications: ELISA/EIA/Western blot.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty