Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,1,1,3,3,3-Hexafluoro-2-propanol-[D₂] (>99,5% D)

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: 1,1,1,3,3,3-Hexafluoroisopropanol-d2HFIP-D21,1,1,3,3,3-Hexafluoro-2-propanol-[²H₂]

DE600-1ML
77401-260EA 223 USD InStock
77401-260
1,1,1,3,3,3-Hexafluoro-2-propanol-[D₂] (>99,5% D)
1,1,1,3,3,3-Hexafluoro-2-propanol-[D2]


Formula: F₃CCD(OD)CF₃
MW: 170.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00037559
CAS Number: 38701-74-5
EINECS: 254-091-9
UN: 3265
ADR: 8,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty