Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ytterbium(III) chloride hydrate 99.9+%

Supplier: APOLLO SCIENTIFIC
Warning

IN3799-50G IN3799-10G
77401-612EA 51 USD InStock
77401-612 77401-614
Ytterbium(III) chloride hydrate 99.9+%
Ytterbium (III) chloride hydrate


Formula: Cl₃Yb·H₂O
MW: 297.41 g/mol
Melting Pt: 150…155 °C
Density: 2.575 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149884
CAS Number: 19423-87-1
EINECS: 233-800-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty