Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium periodate 99%

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: Sodium metaperiodate

IN3306-100G IN3306-500G
77401-562EA 41 USD InStock
77401-562 77401-564
Sodium periodate 99%
Sodium periodate


Formula: NaIO₄
MW: 213.89 g/mol
Melting Pt: 300 °C (dec.)
Density: 4.174 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003534
CAS Number: 7790-28-5
EINECS: 232-197-6
UN: 3085
ADR: 5.1+8,I
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty