Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Strontium oxide 99,5%

Supplier: APOLLO SCIENTIFIC

IN3373-25G IN3373-100G
77401-568EA 483 USD InStock
77401-568 77401-570
Strontium oxide 99,5%
Strontium oxide


Formula: SrO
Melting Pt: 2430 °C
Density: 4.7 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00049558
CAS Number: 1314-11-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty