Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methanol-[D₄] (>99,5% D)

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: Carbinol-D4Methyl alcohol-D4Methanol-[²H₄]Methyl-D3 alcohol-DWood alcohol-D4Methanol-d4Hydroxy methane-D4Tetradeuteromethanol

DE78G-50ML
77401-302EA 112 USD InStock
77401-302
Methanol-[D₄] (>99,5% D)
Methanol-[D4]


Formula: D₃COD
MW: 36.01 g/mol
Boiling Pt: 65.4 °C
Density: 0.888 g/cm³ (25 °C)
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00044637
CAS Number: 811-98-3
EINECS: 212-378-6
UN: 1230
ADR: 3+6.1,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty