Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dichloromethane-[D₂] (>99.50% D)

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: Dichloromethane-[²H₂]Dideuteromethylene chlorideMethylene chloride-D2Methylene chloride-d2Dichloromethane-d2

DE80C-5ML
77401-306EA 51 USD InStock
77401-306
Dichloromethane-[D₂] (>99.50% D)
Dichloromethane-[D2]
(H2O)<0.02%


Formula: CD₂Cl₂
MW: 86.92 g/mol
Boiling Pt: 40 °C
Melting Pt: –97 °C
Density: 1.362 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000882
CAS Number: 1665-00-5
EINECS: 216-776-0
UN: 1593
ADR: 6.1,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty