Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ferric pyrophosphate, Technical Grade

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: Iron(III) pyrophosphateIron (III) pyrophosphate

IN2087-500G
77401-472EA 121 USD InStock
77401-472
Ferric pyrophosphate, Technical Grade
Ferric pyrophosphate


Formula: Fe₄O₂₁P₆
MW: 745.2100 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00016091
CAS Number: 10058-44-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty