Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Zinc oxalate dihydrate 99%

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: Zinc(II) ethane-1,2-dioate dihydrate

IN3886-5G IN3886-25G
77401-620EA 172 USD InStock
77401-620 77401-622
Zinc oxalate dihydrate 99%
Zinc oxalate dihydrate


Formula: ZnC₂O₄·2H₂O
MW: 189.42 g/mol (153.39 anhy)
Melting Pt: 100 °C (dec.)
Density: 2.562 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150611
CAS Number: 55906-21-3
UN: 3288
ADR: 6.1,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty