Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Bromo-5-nitropyridine

Supplier: APOLLO SCIENTIFIC
Danger

OR0936-25G OR0936-100G
77402-322EA 23 USD InStock
77402-322
2-Bromo-5-nitropyridine
2-Bromo-5-nitropyridine


Formula: C₅H₃BrN₂O₂
MW: 203 g/mol
Boiling Pt: 145…147 °C/10 mmHg
Melting Pt: 139…141 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006222
CAS Number: 4487-59-6
EINECS: 224-777-2
UN: 2811
ADR: 6.1,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty