Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Iodobenzene 98%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: Phenyliodide

OR1085-500G OR1085-100G OR1085-25G
77402-956EA 168 USD InStock
77402-956 77402-954 77402-952
Iodobenzene 98%
Iodobenzene


Formula: C₆H₅I
MW: 204.01 g/mol
Boiling Pt: 188…189 °C
Melting Pt: –31 °C
Density: 1.826 g/cm³ (25 °C)
Flash Pt: 74 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001029
CAS Number: 591-50-4
EINECS: 209-719-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty