Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,4-Ethylenedioxythiophene 98%

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: EDOT2,3-Dihydrothieno[3,4-b][1,4]dioxine

OR17830-5G OR17830-100G OR17830-25G
77406-112EA 159 USD InStock
77406-112 77406-116 77406-114
3,4-Ethylenedioxythiophene 98%
3,4-Ethylenedioxythiophene


Formula: C₆H₆O₂S
MW: 142.17 g/mol
Boiling Pt: 193 °C
Melting Pt: 10.5 °C
Density: 1.331 g/cm³ (25 °C)
Flash Pt: 104 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02093622
CAS Number: 126213-50-1
UN: 2810
ADR: 6.1,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty