Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Methoxy-N-methylbenzylamine

Supplier: APOLLO SCIENTIFIC
Danger

OR13981-250MG
77404-778EA 13 USD InStock
77404-778
4-Methoxy-N-methylbenzylamine
4-Methoxy-N-methylbenzylamine


Formula: C₉H₁₃NO
MW: 151.21 g/mol
Boiling Pt: 88…90 °C/1 mmHg
Storage Temperature: Ambient
MDL Number: MFCD04115407
CAS Number: 702-24-9
UN: 2735
ADR: 8,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty