Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

3-Amino-1H-1,2,4-triazole

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 3-Amino-1H-1,2,4-triazoleAmitrole3-Amino-1,2,4-triazole

OR13640-10G OR13640-250G OR13640-50G
77404-700EA 140 USD InStock
77404-700 77404-698 77404-702
3-Amino-1H-1,2,4-triazole
3-Amino-1H-1,2,4-triazole


Formula: C₂H₄N₄
Melting Pt: 149…153 °C
Density: 1.138 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00053362
CAS Number: 61-82-5
EINECS: 200-521-5
UN: 3077
ADR: 9+EHS,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty