Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,2,2-Trichloroethyl chloroformate 95%

Supplier: APOLLO SCIENTIFIC
Danger

OR16412-25G OR16412-500G OR16412-100G
77405-676EA 102 USD InStock
77405-676 77405-678 77405-680
2,2,2-Trichloroethyl chloroformate 95%
2,2,2-Trichloroethyl chloroformate


Formula: C₃H₂Cl₄O₂
MW: 211.86 g/mol
Boiling Pt: 171…172 °C
Melting Pt: 0…2 °C
Density: 1.539 g/cm³ (25 °C)
Flash Pt: >100 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000810
CAS Number: 17341-93-4
EINECS: 241-363-7
UN: 3277
ADR: 6.1+8,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty