Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Iodotrimethylsilane

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: Trimethylsilyl iodideTrimethyliodosilane

OR16935-250G OR16935-100G
77405-854EA 77 USD InStock
77405-854 77405-856
Iodotrimethylsilane
Iodotrimethylsilane


Formula: C₃H₉ISi
MW: 200.09 g/mol
Boiling Pt: 105…106 °C
Density: 1.406 g/cm³ (25 °C)
Flash Pt: –2 °C
Storage Temperature: Freezer
MDL Number: MFCD00001028
CAS Number: 16029-98-4
EINECS: 240-171-0
UN: 2924
ADR: 3+8,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty