Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Nitro-m-toluidine 97%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 3-Methyl-2-nitroaniline

OR16981-1G OR16981-5G OR16981-25G
77405-862EA 23 USD InStock
77405-862 77405-864 77405-866
2-Nitro-m-toluidine 97%
2-Nitro-m-toluidine


Formula: C₇H₈N₂O₂
MW: 152.1500 g/mol
Melting Pt: 105 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00074851
CAS Number: 601-87-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty