Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chloro(dimethyl)(phenyl)silane 95%

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: Chlorodimethylphenylsilane

OR22666-25G OR22666-100G OR22666-250G
77406-928EA 96 USD InStock
77406-928 77406-932 77406-930
Chloro(dimethyl)(phenyl)silane 95%
Chloro(dimethyl)(phenyl)silane


Formula: C₈H₁₁ClSi
MW: 170.71 g/mol
Boiling Pt: 192…193 °C
Density: 1.017 g/cm³ (25 °C)
Flash Pt: 62 °C
Storage Temperature: Ambient
MDL Number: MFCD00000499
CAS Number: 768-33-2
EINECS: 212-193-0
UN: 2987
ADR: 8,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty