Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Methyl-4-phenyl-1,2,3-thiadiazole 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR26197-5G OR26197-250MG OR26197-1G
77407-760EA 254 USD InStock
77407-760 77407-758 77407-756
5-Methyl-4-phenyl-1,2,3-thiadiazole 95%
5-Methyl-4-phenyl-1,2,3-thiadiazole


Formula: C₉H₈N₂S
Storage Temperature: Ambient
MDL Number: MFCD00100201
CAS Number: 64273-28-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty