Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

α-Chloro-o-xylene 98+%

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: 2-Methylbenzyl chloride

OR28627-1KG OR28627-5KG OR28627-500G
77408-132EA 252 USD InStock
77408-132 77408-130 77408-128
α-Chloro-o-xylene 98+%
α-Chloro-o-xylene
Intermediate for organic synthesis.


Formula: C₈H₉Cl
MW: 140.61 g/mol
Boiling Pt: 197…199 °C
Melting Pt: 2 °C
Density: 1.063 g/cm³ (25 °C)
Flash Pt: 73.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000899
CAS Number: 552-45-4
EINECS: 209-013-8
UN: 3265
ADR: 8,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty