Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Butyric acid 95%

Supplier: APOLLO SCIENTIFIC
Synonyms: n-Butyric acidButanoic acid

OR6005-100G
77410-666EA 89 USD InStock
77410-666
Butyric acid 95%
Butyric acid


Formula: C₄H₈O₂
MW: 88.11 g/mol
Boiling Pt: 162 °C
Melting Pt: –12 °C
Density: 0.964 g/cm³ (25 °C)
Flash Pt: 72 °C
Storage Temperature: Ambient
MDL Number: MFCD00002814
CAS Number: 107-92-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty