Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Mercapto-1,2-propanediol 97%

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: (±)-3-Mercapto-1,2-dihydroxy propane(±)-3-Mercapto-1,2-propanediol(±)-α-Monothioglycerinealpha-Thioglycerolα-Monothioglycerol(±)-1-Thioglycerine1-ThioglycerolMonothioglycerol

OR61438-10ML
77410-748EA 20 USD InStock
77410-748
3-Mercapto-1,2-propanediol 97%
3-Mercapto-1,2-propanediol


Formula: C₃H₈O₂S
MW: 108.16 g/mol
Boiling Pt: 118 °C/5 mmHg
Density: 1.247 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004879
CAS Number: 96-27-5
EINECS: 202-495-0
UN: 2810
ADR: 6.1,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty