Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L-Phenylalaninol 97%

Supplier: APOLLO SCIENTIFIC
Synonyms: L(-)-2-Amino-3-phenylpropan-1-ol(S)-(-)-Phenylalaninol(S)-2-Amino-3-phenylpropan-1-olL(-)-2-Amino-3-phenyl-1-propanolH-Phenylalaninol(S)-(-)-2-Amino-3-phenyl-1-propanol(S)-(-)-2-Amino-3-phenylpropan-1-olL(-)-Phenylalaninol

OR5852-100G
77410-546EA 30 USD InStock
77410-546
L-Phenylalaninol 97%
L-Phenylalaninol


Formula: C₉H₁₃NO
MW: 151.21 g/mol
Melting Pt: 92…94 °C
Storage Temperature: Ambient
MDL Number: MFCD00004732
CAS Number: 3182-95-4
UN: 3259
ADR: 8,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty