Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,4,6-Trimethylbenzaldehyde 95%

Supplier: APOLLO SCIENTIFIC
Synonyms: Mesitaldehyde

OR911451-100G OR911451-500G
77411-612EA 102 USD InStock
77411-612 77411-614
2,4,6-Trimethylbenzaldehyde 95%
2,4,6-Trimethylbenzaldehyde


Formula: C₁₀H₁₂O
MW: 148.2 g/mol
Boiling Pt: 237 °C/760 mmHg
Melting Pt: 11 °C
Density: 1.02 g/cm³ (25 °C)
Flash Pt: 104 °C
Storage Temperature: Ambient
MDL Number: MFCD00003341
CAS Number: 487-68-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty