Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-(2-Bromoethyl)-1,3-dioxolane

Supplier: APOLLO SCIENTIFIC
Danger

OR52533-5G
77410-254EA 13 USD InStock
77410-254
2-(2-Bromoethyl)-1,3-dioxolane
2-(2-Bromoethyl)-1,3-dioxolane


Formula: C₅H₉BrO₂
Boiling Pt: 68…70 °C/8 mmHg
Density: 1.542 g/cm³ (25 °C)
Flash Pt: 65 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003216
CAS Number: 18742-02-4
EINECS: 242-551-1
UN: 2810
ADR: 6.1,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty