Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Genipin 98%

Supplier: APOLLO SCIENTIFIC
Synonyms: Methyl (1S,2R,6S)-2-Hydroxy-9-(hydroxymethyl)-3-oxabicyclo[4.3.0]nona-4,8-diene-5-carboxylate(1R,4aS,7aS)-Methyl 1-hydroxy-7-(hydroxymethyl)-1,4a,5,7a-tetrahydrocyclopenta[c]pyran-4-carboxylate

OR932863-5G OR932863-1G
77411-992EA 21 USD InStock
77411-992 77411-994
Genipin 98%
Genipin


Formula: C₁₁H₁₄O₅
MW: 226,23 g/mol
Melting Pt: 120…121 °C
Storage Temperature: Ambient
MDL Number: MFCD00888600
CAS Number: 6902-77-8
UN: 2811
ADR: 6.1,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty