Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-(3-Bromophenyl)-3-methyl-1,2,4-oxadiazole 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR9301-5G OR9301-1G OR9301-250MG
77411-938EA 220 USD InStock
77411-938 77411-934 77411-936
5-(3-Bromophenyl)-3-methyl-1,2,4-oxadiazole 97%
5-(3-Bromophenyl)-3-methyl-1,2,4-oxadiazole


Formula: C₉H₇BrN₂O
Melting Pt: 95…96 °C
Storage Temperature: Ambient
MDL Number: MFCD08435902
CAS Number: 160377-58-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty