Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium isopropylxanthate 95%

Supplier: APOLLO SCIENTIFIC
Synonyms: Isopropylxanthic Acid Potassium Salt

OR938096-25G OR938096-5G
77412-024EA 40 USD InStock
77412-024 77412-022
Potassium isopropylxanthate 95%
Potassium isopropylxanthate


Formula: C₄H₈OS₂
MW: 174.32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004930
CAS Number: 140-92-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty