Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Bromo-5-(trifluoromethyl)pyridine 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC1598G-5G PC1598G-25G PC1598G-1G
77413-252EA 41 USD InStock
77413-252 77413-254 77413-250
2-Bromo-5-(trifluoromethyl)pyridine 97%
2-Bromo-5-(trifluoromethyl)pyridine


Formula: C₆H₃BrF₃N
MW: 226 g/mol
Boiling Pt: 78…81 °C/30 mmHg
Melting Pt: 44…48 °C
Storage Temperature: Ambient
MDL Number: MFCD00153086
CAS Number: 50488-42-1
UN: 2811
ADR: 6.1,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty