Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(Pentafluoroethyl)trimethylsilane 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC1217-25G PC1217-1G PC1217-5G
77413-088EA 214 USD InStock
77413-088 77413-090 77413-086
(Pentafluoroethyl)trimethylsilane 97%
(Pentafluoroethyl)trimethylsilane


Formula: C₅H₉F₅Si
Boiling Pt: 60…70 °C
Density: 1.11 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD04116436
CAS Number: 124898-13-1
UN: 1993
ADR: 3,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty