Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Fluorophenyl isocyanate 98%

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: 4-Fluorophenylisocyanate1-Fluoro-4-isocyanatobenzene

PC4166-100G PC4166-25G PC4166-5G
77415-182EA 111 USD InStock
77415-182 77415-186 77415-184
4-Fluorophenyl isocyanate 98%
4-Fluorophenyl isocyanate
  • May form small amount of sediment.


Formula: C₇H₄FNO
MW: 137.11 g/mol
Boiling Pt: 55 °C/8 mmHg
Density: 1.206 g/cm³ (25 °C)
Flash Pt: 52 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002023
CAS Number: 1195-45-5
EINECS: 214-799-0
UN: 3080
ADR: 6.1+3,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty