Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl-3-(trifluoromethyl)crotonate 97%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 3-(Trifluoromethyl)crotonic acid ethyl ester

PC3288C-25G PC3288C-5G
77414-660EA 350 USD InStock
77414-660 77414-662
Ethyl-3-(trifluoromethyl)crotonate 97%
Ethyl-3-(trifluoromethyl)crotonate


Formula: C₇H₉F₃O₂
MW: 182.14
Boiling Pt: 129…130 °C
Flash Pt: 32 °C
Storage Temperature: Ambient
MDL Number: MFCD00040846
CAS Number: 24490-03-7
UN: 3272
ADR: 3,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty