Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl-3-(trifluoromethyl)benzoate 95%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: Methyl 3-(trifluoromethyl)benzoate

PC5171Z-500G PC5171Z-100G PC5171Z-25G
77416-456EA 13 USD InStock
77416-456 77416-458 77416-454
Methyl-3-(trifluoromethyl)benzoate 95%
Methyl 3-(trifluoromethyl)benzoate


Formula: C₉H₇F₃O₂
MW: 204.15 g/mol
Boiling Pt: 93…95 °C/20 mmHg
Density: 1.295 g/cm³ (25 °C)
Flash Pt: 40 °C
Storage Temperature: Ambient
MDL Number: MFCD00043543
CAS Number: 2557-13-3
UN: 3272
ADR: 3,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty