Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(4-(Trifluoromethyl)phenyl)methanesulfonyl chloride

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: 4-Trifluoromethyl-α-toluenesulphonyl chloride[4-(Trifluoromethyl)phenyl]methanesulphonyl chloride[4-(Trifluoromethyl)phenyl]methanesulfonyl chloride4-Trifluoromethyl-α-toluenesulfonyl chloride

PC49173-250MG
77415-876EA 13 USD InStock
77415-876
(4-(Trifluoromethyl)phenyl)methanesulfonyl chloride
(4-(Trifluoromethyl)phenyl)methanesulphonyl chloride


Formula: C₈H₆ClF₃O₂S
MW: 258.65 g/mol
Melting Pt: 103…107 °C
Storage Temperature: Ambient
UN: 3261
ADR: 8,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty