Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lithium fluoride 99.99%

Supplier: APOLLO SCIENTIFIC
Danger

PC4990R-50G PC4990R-250G
77415-978EA 506 USD InStock
77415-978 77415-980
Lithium fluoride 99.99%
Lithium fluoride


Formula: FLi
MW: 25.94 g/mol
Boiling Pt: 1680 °C
Melting Pt: 845 °C
Density: 2.635 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011090
CAS Number: 7789-24-4
EINECS: 232-152-0
UN: 3288
ADR: 6.1,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty