Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,3,4,5-Tetrafluorobenzoyl chloride 98%

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: 2,3,4,5-tetrafluorobenzoylchloride

PC6742-100G PC6742-500G PC6742-25G
77417-526EA 13 USD InStock
77417-526 77417-524 77417-528
2,3,4,5-Tetrafluorobenzoyl chloride 98%
2,3,4,5-Tetrafluorobenzoyl chloride


Formula: C₇HClF₄O
MW: 212.53 g/mol
Density: 1.58 g/cm³ (25 °C)
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00075164
CAS Number: 94695-48-4
UN: 3265
ADR: 8,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty